Learn More
Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%, Thermo Scientific Chemicals
£74.20 - £277.00
Chemical Identifiers
CAS | 14337-43-0 |
---|---|
Molecular Formula | C4H6ClNO3 |
Molecular Weight (g/mol) | 151.55 |
MDL Number | MFCD00010209 |
InChI Key | UXOLDCOJRAMLTQ-UTCJRWHESA-N |
Synonym | ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate |
PubChem CID | 9576005 |
IUPAC Name | ethyl (2Z)-2-chloro-2-hydroxyiminoacetate |
SMILES | CCOC(=O)C(\Cl)=N\O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10366924
|
Thermo Scientific Acros
398740050 |
5 g | Glass bottle |
£74.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10182854
|
Thermo Scientific Acros
398740250 |
25 g | Glass bottle |
£277.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14337-43-0 | |
151.55 | |
UXOLDCOJRAMLTQ-UTCJRWHESA-N | |
9576005 | |
CCOC(=O)C(\Cl)=N\O |
C4H6ClNO3 | |
MFCD00010209 | |
ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate | |
ethyl (2Z)-2-chloro-2-hydroxyiminoacetate |
Specifications
14337-43-0 | |
96% min. (Argentometry) | |
MFCD00010209 | |
UXOLDCOJRAMLTQ-UTCJRWHESA-N | |
ethyl (2Z)-2-chloro-2-hydroxyiminoacetate | |
9576005 | |
97% |
70°C to 76°C | |
C4H6ClNO3 | |
ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate | |
CCOC(=O)C(\Cl)=N\O | |
151.55 | |
151.55 | |
Ethyl 2-chloro-2-(hydroxyimino)acetate |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specificatio
GHS Signal Word: Danger
- Ethyl 2-chloro-2-(hydroxyimino)acetate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only