missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate, 95%, Thermo Scientific™
£26.32 - £75.60
Chemical Identifiers
CAS | 861208-16-4 |
---|---|
Molecular Formula | C10H8BrClN2O2 |
Molecular Weight (g/mol) | 303.54 |
MDL Number | MFCD06496148 |
InChI Key | PEINUXVIHDZXNP-UHFFFAOYSA-N |
Synonym | ethyl 3-bromo-6-chloroimidazo 1,2-a pyridine-2-carboxylate, ethyl 3-bromo-6-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylate, 3-bromo-6-chloro-imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester hydrobromide |
PubChem CID | 5260614 |
IUPAC Name | ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate |
SMILES | CCOC(=O)C1=C(Br)N2C=C(Cl)C=CC2=N1 |
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische Identifikatoren
861208-16-4 | |
303.54 | |
PEINUXVIHDZXNP-UHFFFAOYSA-N | |
5260614 | |
CCOC(=O)C1=C(Br)N2C=C(Cl)C=CC2=N1 |
C10H8BrClN2O2 | |
MFCD06496148 | |
ethyl 3-bromo-6-chloroimidazo 1,2-a pyridine-2-carboxylate, ethyl 3-bromo-6-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylate, 3-bromo-6-chloro-imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester hydrobromide | |
ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate |
Spezifikation
861208-16-4 | |
C10H8BrClN2O2 | |
ethyl 3-bromo-6-chloroimidazo 1,2-a pyridine-2-carboxylate, ethyl 3-bromo-6-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylate, 3-bromo-6-chloro-imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester hydrobromide | |
CCOC(=O)C1=C(Br)N2C=C(Cl)C=CC2=N1 | |
303.54 | |
303.54 | |
Ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate |
139°C to 141°C | |
MFCD06496148 | |
PEINUXVIHDZXNP-UHFFFAOYSA-N | |
ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate | |
5260614 | |
95% |
Sicherheit und Handhabung
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures