missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 3-cyano-3-phenylpyruvate, 97%, Thermo Scientific Chemicals Product Code.: 11451550

Ethyl 3-cyano-3-phenylpyruvate, 97%, Thermo Scientific Chemicals

Product Code. 11451550
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11451550

Brand: Thermo Scientific Alfa Aesar H26951.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6362-63-6
Molecular Formula C12H11NO3
Molecular Weight (g/mol) 217.22
MDL Number MFCD00019801
InChI Key YWNRATDDUSMBPR-UHFFFAOYNA-N
Synonym ethyl 3-cyano-3-phenylpyruvate, ethyl phenylcyanopyruvate, cyanophenylpyruvic acid ethyl ester, ethyl cyanophenylpyruvate, ethyl alpha-cyanophenylpyruvate, pyruvic acid, cyanophenyl-, ethyl ester, ethyl .alpha.-cyanophenylpyruvate, wln: ncr x1v1uno2, 4-10-00-03440 beilstein handbook reference, benzenepropanoic acid, .beta.-cyano-.alpha.-oxo-, ethyl esterShow MoreShow Less
PubChem CID 22845
IUPAC Name ethyl 3-cyano-2-oxo-3-phenylpropanoate
SMILES CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1

Specifications

Melting Point 125°C to 129°C
Quantity 25 g
Formula Weight 217.23
Percent Purity 97%
Chemical Name or Material Ethyl 3-cyano-3-phenylpyruvate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10240642 Certificate TypeCertificate of Analysis Date25/05/2022 Catalog NumberH26951.14,H26951.06
Lot Number10136479 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH26951.14,H26951.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3-cyano-3-phenylpyruvate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.