Learn More
Ethyl 3-iodobenzoate, 99%, Thermo Scientific Chemicals
£45.10 - £513.00
Chemical Identifiers
CAS | 58313-23-8 |
---|---|
Molecular Formula | C9H9IO2 |
Molecular Weight (g/mol) | 276.073 |
MDL Number | MFCD00044769 |
InChI Key | POGCXCWRMMXDAQ-UHFFFAOYSA-N |
Synonym | ethyl-3-iodobenzoate, 3-iodobenzoic acid ethyl ester, benzoic acid, 3-iodo-, ethyl ester, ethyl m-iodobenzoate, m-iodobenzoic acid ethyl ester, pubchem3883, ethyl-3-iodo-benzoate, acmc-1aupw, ethyl 3-iodobenzoate, rarechem al bi 0709 |
PubChem CID | 143542 |
IUPAC Name | ethyl 3-iodobenzoate |
SMILES | CCOC(=O)C1=CC(=CC=C1)I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11442297
|
Thermo Scientific Alfa Aesar
A14210.09 |
10 g | N/A | ||||||
11452297
|
Thermo Scientific Alfa Aesar
A14210.18 |
50 g |
£154.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11462297
|
Thermo Scientific Alfa Aesar
A14210.30 |
250 g | N/A | ||||||
Description
Ethyl 3-iodobenzoate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 3-iodobenzoate is used as pharmaceutical intermediates.
Solubility
Not miscible in water.
Notes
Light Sensitive. Store in the dark. Store away from oxidizing agents.
Chemical Identifiers
58313-23-8 | |
276.073 | |
POGCXCWRMMXDAQ-UHFFFAOYSA-N | |
143542 | |
CCOC(=O)C1=CC(=CC=C1)I |
C9H9IO2 | |
MFCD00044769 | |
ethyl-3-iodobenzoate, 3-iodobenzoic acid ethyl ester, benzoic acid, 3-iodo-, ethyl ester, ethyl m-iodobenzoate, m-iodobenzoic acid ethyl ester, pubchem3883, ethyl-3-iodo-benzoate, acmc-1aupw, ethyl 3-iodobenzoate, rarechem al bi 0709 | |
ethyl 3-iodobenzoate |
Specifications
58313-23-8 | |
272°C | |
C9H9IO2 | |
MFCD00044769 | |
Light sensitive | |
Not miscible in water. | |
CCOC(=O)C1=CC(=CC=C1)I | |
276.073 | |
276.08 | |
Ethyl 3-iodobenzoate |
1.66 | |
>110°C (230°F) | |
1.5815 | |
2089064 | |
ethyl-3-iodobenzoate, 3-iodobenzoic acid ethyl ester, benzoic acid, 3-iodo-, ethyl ester, ethyl m-iodobenzoate, m-iodobenzoic acid ethyl ester, pubchem3883, ethyl-3-iodo-benzoate, acmc-1aupw, ethyl 3-iodobenzoate, rarechem al bi 0709 | |
POGCXCWRMMXDAQ-UHFFFAOYSA-N | |
ethyl 3-iodobenzoate | |
143542 | |
99% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only