missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-oxo-3-(4-pyridyl)propionate, 98%, Thermo Scientific Chemicals
£58.00 - £293.00
Chemical Identifiers
CAS | 26377-17-3 |
---|---|
Molecular Formula | C10H11NO3 |
Molecular Weight (g/mol) | 193.20 |
MDL Number | MFCD00094021 |
InChI Key | PCJNYGPKMQQCPX-UHFFFAOYSA-N |
Synonym | ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester |
PubChem CID | 2735202 |
IUPAC Name | ethyl 3-oxo-3-pyridin-4-ylpropanoate |
SMILES | CCOC(=O)CC(=O)C1=CC=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478208
|
Thermo Scientific Alfa Aesar
H61647.06 |
5 g |
£58.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15488208
|
Thermo Scientific Alfa Aesar
H61647.14 |
25 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26377-17-3 | |
193.20 | |
PCJNYGPKMQQCPX-UHFFFAOYSA-N | |
2735202 | |
CCOC(=O)CC(=O)C1=CC=NC=C1 |
C10H11NO3 | |
MFCD00094021 | |
ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester | |
ethyl 3-oxo-3-pyridin-4-ylpropanoate |
Specifications
26377-17-3 | |
Brown | |
C10H11NO3 | |
MFCD00094021 | |
PCJNYGPKMQQCPX-UHFFFAOYSA-N | |
ethyl 3-oxo-3-pyridin-4-ylpropanoate | |
2735202 | |
98% | |
Ethyl 3-oxo-3-(4-pyridyl)propionate |
53°C to 59°C | |
1.155 g/mL | |
1.51 | |
ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester | |
CCOC(=O)CC(=O)C1=CC=NC=C1 | |
193.20 | |
193.2 | |
Solid |
Safety and Handling
missing translation for 'einecsNumber' : 247-635-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only