missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (4-fluorobenzoyl)acetate, 95%, Thermo Scientific Chemicals
£18.90 - £200.00
Chemical Identifiers
CAS | 1999-00-4 |
---|---|
Molecular Formula | C11H11FO3 |
Molecular Weight (g/mol) | 210.204 |
MDL Number | MFCD03093631 |
InChI Key | SJUXLKYJKQBZLM-UHFFFAOYSA-N |
Synonym | ethyl 3-4-fluorophenyl-3-oxopropanoate, ethyl 4-fluorobenzoylacetate, ethyl 4-fluorobenzoyl acetate, 4-fluorobenzoylacetic acid ethyl ester, ethyl p-fluorobenzoyl acetate, 4-fluorobenzoyl acetic acid ethyl ester, ethyl 3-4-fluorophenyl-3-oxopropionate, ethyl 3-4'-fluorophenyl-3-oxopropanoate, ethyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid ethyl ester |
PubChem CID | 2758844 |
IUPAC Name | ethyl 3-(4-fluorophenyl)-3-oxopropanoate |
SMILES | CCOC(=O)CC(=O)C1=CC=C(C=C1)F |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1999-00-4 | |
210.204 | |
SJUXLKYJKQBZLM-UHFFFAOYSA-N | |
2758844 | |
CCOC(=O)CC(=O)C1=CC=C(C=C1)F |
C11H11FO3 | |
MFCD03093631 | |
ethyl 3-4-fluorophenyl-3-oxopropanoate, ethyl 4-fluorobenzoylacetate, ethyl 4-fluorobenzoyl acetate, 4-fluorobenzoylacetic acid ethyl ester, ethyl p-fluorobenzoyl acetate, 4-fluorobenzoyl acetic acid ethyl ester, ethyl 3-4-fluorophenyl-3-oxopropionate, ethyl 3-4'-fluorophenyl-3-oxopropanoate, ethyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid ethyl ester | |
ethyl 3-(4-fluorophenyl)-3-oxopropanoate |
Specifications
1999-00-4 | |
1.174 | |
1.504 | |
ethyl 3-4-fluorophenyl-3-oxopropanoate, ethyl 4-fluorobenzoylacetate, ethyl 4-fluorobenzoyl acetate, 4-fluorobenzoylacetic acid ethyl ester, ethyl p-fluorobenzoyl acetate, 4-fluorobenzoyl acetic acid ethyl ester, ethyl 3-4-fluorophenyl-3-oxopropionate, ethyl 3-4'-fluorophenyl-3-oxopropanoate, ethyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid ethyl ester | |
CCOC(=O)CC(=O)C1=CC=C(C=C1)F | |
210.204 | |
210.2 | |
Ethyl (4-fluorobenzoyl)acetate |
117°C to 120°C | |
C11H11FO3 | |
MFCD03093631 | |
SJUXLKYJKQBZLM-UHFFFAOYSA-N | |
ethyl 3-(4-fluorophenyl)-3-oxopropanoate | |
2758844 | |
95% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only