missing translation for 'onlineSavingsMsg'
Learn More
Ethyl chloroformate, 97%, Thermo Scientific Chemicals Product Code.: 10167613

Ethyl chloroformate, 97%, Thermo Scientific Chemicals

Product Code. 10167613
100 mL, Glass bottle
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10167613

Brand: Thermo Scientific Acros 181081000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-41-3
InChI Key RIFGWPKJUGCATF-UHFFFAOYSA-N
Synonym ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiatoShow MoreShow Less
PubChem CID 10928
IUPAC Name ethyl carbonochloridate
SMILES CCOC(=O)Cl

Specifications

Density 1.1350g/mL
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C3H5ClO2
Linear Formula ClCO2C2H5
Quantity 100 mL
Fieser 01,364; 02,193; 04,228; 05,294; 07,147; 09,214; 12,223
Molecular Weight (g/mol) 108.521
Formula Weight 108.52
Chemical Name or Material Ethyl chloroformate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465513 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number181081000,181080025,181080050,181085000
Lot NumberA0463401 Certificate TypeCertificate of Analysis Date06/06/2024 Catalog Number181081000,181080025,181080050,181085000
Lot NumberA0460991 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog Number181081000,181080025,181080050,181085000
Lot NumberA0442831 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number181081000,181080025,181080050,181085000
Lot NumberA0394839 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number181081000,181080025,181080050,181085000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl chloroformate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.