Learn More
Masterflex™ Ethyl Vinyl Acetate Microbore Tubing
Use this tubing for biological testing. Saint Gobain Performance Plastics™ Ethyl Vinyl Acetate Microbore Tubing is biologically stable, nontoxic and non-contaminating.
£72.60 - £287.00
Specifications
Material EVA
Packaging Quantity 1 Pack
View More Specs
Products 9
Product Code Brand Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity & Availability  
11977875
View Documents Promotion Details
Masterflex™
95712-12
0.25 mm 2.08 mm
£75.60
15m
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11727355
View Documents Promotion Details
Masterflex™
06492-02
0.51 mm 1.52 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11917885
View Documents Promotion Details
Masterflex™
95712-30
1.02 mm 3.07 mm
£77.35
15m
View alternative products
Save Up to 
»
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
11787355
View Documents Promotion Details
Masterflex™
06492-03
0.76 mm 2.29 mm
£287.00
30m
View alternative products
Save Up to 
»
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
11707365
View Documents Promotion Details
Masterflex™
06492-05
1.27 mm 2.29 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11927885
View Documents Promotion Details
Masterflex™
95712-32
1.27 mm 3.23 mm
£72.60
15m
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11937885
View Documents Promotion Details
Masterflex™
95712-34
1.42 mm 3.35 mm
£72.60
15m
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
11947885
View Documents Promotion Details
Masterflex™
95712-36
1.52 mm 3.45 mm
£77.35
15m
View alternative products
Save Up to 
»
Estimated Shipment: 13-11-2024
to see stock.
Add to basket
11957885
missing translation for 'viewDocuments' Promotion Details
Masterflex™
95712-40
1.85 mm 3.78 mm
£72.60
15m
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
Description
Description
  • Translucent cross-linked tubing flexible micro-line tubing
  • Virtually 100% elastic memory, returns to its original shape with gentle heating
  • Resin is USP 23 Class VI and National Formulary Vol. 14 specifications
  • Sterilize with autoclave, ethylene oxide, gamma irradiation or dry heat
Specifications
Specifications
EVA
1 Pack