missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene, 99.8%, pure, Thermo Scientific Chemicals

£17.99 - £236.00

Chemical Identifiers

CAS 100-41-4
MDL Number MFCD00011647
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
Synonym phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
PubChem CID 7500
ChEBI CHEBI:16101
IUPAC Name ethylbenzene
SMILES CCC1=CC=CC=C1
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10687642
View Documents
Thermo Scientific Acros
118080250
25 mL Glass bottle
£17.99
25mL
Please to purchase this item. Need a web account? Register with us today!
10448472
View Documents
Thermo Scientific Acros
118085000
500 mL Glass bottle
£24.62
500mL
Please to purchase this item. Need a web account? Register with us today!
15871022
View Documents
Thermo Scientific Acros
118080010
1 L Glass bottle
£28.14
1 L
Please to purchase this item. Need a web account? Register with us today!
10589420
View Documents
Thermo Scientific Acros
118080025
2.5 L Glass bottle
£34.47
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10339444
View Documents Promotion Details
Thermo Scientific Acros
118080251
25 L Metal Drum
£236.00
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100-41-4
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
7500
ethylbenzene
MFCD00011647
phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
CHEBI:16101
CCC1=CC=CC=C1

Specifications

100-41-4
0.8600g/mL
22°C
99.7% min. (GC)
C6H5C2H5
MFCD00011647
15, 3820
phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
ethylbenzene
7500
0.63 mPa.s (20°C)
99.8%
Liquid
-95.0°C
136.0°C
Authentic
C8H10
1.4940 to 1.4970
05, II, 274
0.86
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, benzene, carbon tetrachloride,, ether and sulfur dioxide, slightly soluble in chloroform, insoluble in ammonia
CCC1=CC=CC=C1
106.17
CHEBI:16101
106.17
Pure
Ethylbenzene, 99.8%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasti

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-849-4

missing translation for 'rtecsNumber' : DA0700000

missing translation for 'tsca' : TSCA

Product Identifier
  • Ethylbenzene

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H304-May be fatal if swallowed and enters airways.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.