missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
C10H18N2Na2O10, CAS Number-6381-92-6, 5kg, White, 372.23g/mol, 252 deg.C, 150037, Plastic keg, 4 to 6, Solid

Specifications
Specifications
Melting Point | 252°C |
Color | White |
pH | 4 to 6 |
Physical Form | Solid |
Quantity | 100 g |
Formula Weight | 372.23g/mol |
Packaging | Plastic powder jar |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 15
Safety and Handling


Product Identifier
- Ethylenediaminetetraacetic acid, disodium salt dihydrate
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- P304-IF INHALED:Call a POISON CENTER/doctor if you feelunwell..,IF INHALED:Call a POISON CENTER/doctor if you feelunwell..
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.,Avoid breathing dust/fume/gas/mist/vapours/spray.
- MIXTURE LIST-Contains : Ethylenediaminetetraacetic acid, disodium salt dihydrate, Disodium EDTA
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Certified AR for Analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction