missing translation for 'onlineSavingsMsg'
Learn More
Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals Product Code.: 11389359

Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals

Product Code. 11389359
500 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11389359

Brand: Thermo Scientific Alfa Aesar 088995.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethylenediaminetetraacetic acid, iron(III) monosodium salt is used to eliminate enzyme inhibition by traces of heavy metals and to inhibit enzymes that require divalent cations as cofactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15708-41-5
Molecular Formula C10H12FeN2NaO8
Molecular Weight (g/mol) 367.047
MDL Number MFCD00078215
InChI Key MKWYFZFMAMBPQK-UHFFFAOYSA-J
Synonym sodium feredetate, edta ferric sodium salt, calmosine, sodium ironedetate, edta iron iii sodium salt, ferisan, sytron, sequestrene nafe, sodium iron edta, ferric sodium edtaShow MoreShow Less
PubChem CID 27461
ChEBI CHEBI:78292
IUPAC Name sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+)
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3]

Specifications

Odor Odorless
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 367.05
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, iron (III) monosodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY03C030 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog Number088995.A3,088995.22,088995.36
Lot NumberM12K031 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number088995.A3,088995.22,088995.36
Lot NumberQ26I028 Certificate TypeCertificate of Analysis Date02/05/2022 Catalog Number088995.A3,088995.22,088995.36
Lot NumberG26Y003 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088995.A3,088995.22,088995.36
Lot NumberL12S036 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088995.A3,088995.22,088995.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.