missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylidene diacetate, 99%, Thermo Scientific Chemicals
£37.20 - £400.95
Chemical Identifiers
CAS | 542-10-9 |
---|---|
Molecular Formula | C6H10O4 |
Molecular Weight (g/mol) | 146.14 |
InChI Key | ACKALUBLCWJVNB-UHFFFAOYSA-N |
Synonym | ethylidene diacetate, 1,1-diacetoxyethane, 1,1-ethanediol diacetate, 1,1-ethanediol, diacetate, ethylidene acetate, ethane-1,1-diyl diacetate, polyoxymethylenes, delrin, unii-kl1s8v6w25, 1-acetoxyethyl acetate |
PubChem CID | 222536 |
IUPAC Name | 1-acetyloxyethyl acetate |
SMILES | CC(OC(=O)C)OC(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10137280
|
Thermo Scientific Acros
410130250 |
25 g | Glass bottle | N/A | |||||
10197370
|
Thermo Scientific Acros
410131000 |
100 g | Glass bottle | N/A | |||||
11914921
|
Thermo Scientific Acros
410135000 |
500 g | Glass Bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
542-10-9 | |
146.14 | |
ethylidene diacetate, 1,1-diacetoxyethane, 1,1-ethanediol diacetate, 1,1-ethanediol, diacetate, ethylidene acetate, ethane-1,1-diyl diacetate, polyoxymethylenes, delrin, unii-kl1s8v6w25, 1-acetoxyethyl acetate | |
1-acetyloxyethyl acetate |
C6H10O4 | |
ACKALUBLCWJVNB-UHFFFAOYSA-N | |
222536 | |
CC(OC(=O)C)OC(=O)C |
Specifications
542-10-9 | |
69.0°C (17.0 mmHg) | |
Authentic | |
C6H10O4 | |
1.3995 to 1.4015 | |
ethylidene diacetate, 1,1-diacetoxyethane, 1,1-ethanediol diacetate, 1,1-ethanediol, diacetate, ethylidene acetate, ethane-1,1-diyl diacetate, polyoxymethylenes, delrin, unii-kl1s8v6w25, 1-acetoxyethyl acetate | |
CC(OC(=O)C)OC(=O)C | |
146.14 | |
146.14 | |
Liquid |
Colorless to Yellow | |
68°C | |
98.5% min. (GC) | |
CH3CH(OCOCH3)2 | |
15,3866 | |
ACKALUBLCWJVNB-UHFFFAOYSA-N | |
1-acetyloxyethyl acetate | |
222536 | |
99% | |
Ethylidene diacetate |
Safety and Handling
missing translation for 'einecsNumber' : 208-800-3
Product Identifier
- Ethylidene diacetate
RUO – Research Use Only