Learn More
Eugenol, 99%, Thermo Scientific Chemicals
£23.01 - £421.10
Chemical Identifiers
CAS | 97-53-0 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.20 |
MDL Number | MFCD00008654 |
InChI Key | RRAFCDWBNXTKKO-UHFFFAOYSA-N |
Synonym | eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol |
PubChem CID | 3314 |
ChEBI | CHEBI:4917 |
SMILES | COC1=CC(CC=C)=CC=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10050990
|
Thermo Scientific Acros
119110050 |
5 g | Glass bottle |
£23.01
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10607652
|
Thermo Scientific Acros
119111000 |
100 g | Glass bottle |
£33.37
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10550591
|
Thermo Scientific Acros
119115000 |
500 g | Glass bottle |
£109.55
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11934751
|
Thermo Scientific Acros
119110025 |
2.5 kg | Glass bottle |
£421.10
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97-53-0 | |
164.20 | |
RRAFCDWBNXTKKO-UHFFFAOYSA-N | |
3314 | |
COC1=CC(CC=C)=CC=C1O |
C10H12O2 | |
MFCD00008654 | |
eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol | |
CHEBI:4917 |
Specifications
97-53-0 | |
Yellow | |
254.0°C | |
Authentic | |
C10H12O2 | |
1.5400 to 1.5420 | |
06,961 | |
1.06 | |
Solubility in water: 2.46g/L (25°C). Other solubilities: soluble in glacial acetic acid,soluble in alcohol,chloroform and ether | |
COC1=CC(CC=C)=CC=C1O | |
164.20 | |
CHEBI:4917 | |
99% | |
Eugenol, 99% |
-12°C to 10°C | |
1.0600g/mL | |
>112°C | |
98.5% min. (GC) | |
(H2C=CHCH2)C6H3(OCH3)OH | |
MFCD00008654 | |
15,394 | |
eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol | |
RRAFCDWBNXTKKO-UHFFFAOYSA-N | |
2-methoxy-4-prop-2-enylphenol | |
3314 | |
164.2 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-589-1
missing translation for 'rtecsNumber' : SJ4375000
missing translation for 'tsca' : TSCA
- Eugenol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.