missing translation for 'onlineSavingsMsg'
Learn More

Europium(II) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11338898
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
14077-39-5
EuF2
189.961
MFCD01866333
YLDGIMHVWSTRML-UHFFFAOYSA-L
europium ii fluoride, acmc-20akvn, europium ii fluoride, anhydrous
21225127
europium(2+);difluoride
[F-].[F-].[Eu+2]
This item is not returnable. View return policy
14077-39-5
EuF2
189.961
MFCD01866333
YLDGIMHVWSTRML-UHFFFAOYSA-L
europium ii fluoride, acmc-20akvn, europium ii fluoride, anhydrous
21225127
europium(2+);difluoride
[F-].[F-].[Eu+2]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point >2400°C
Melting Point 1380°C
Physical Form Powder
Quantity 0.5 g
UN Number UN3288
Solubility Information Insoluble in water.
Formula Weight 189.96
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.495 g/mL
Assay (metals basis)
Chemical Name or Material Europium(II) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'