missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals
£128.00 - £356.00
Chemical Identifiers
CAS | 181266-82-0 |
---|---|
Molecular Formula | C15H21EuO6 |
Molecular Weight (g/mol) | 449.29 |
MDL Number | MFCD00192170 |
InChI Key | YMTKZYSHNNXTIJ-UHFFFAOYSA-N |
Synonym | europium iii 2,4-pentanedionate hydrate reo |
PubChem CID | 131675702 |
SMILES | [Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366658
|
Thermo Scientific Alfa Aesar
013209.04 |
2 g |
£128.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376658
|
Thermo Scientific Alfa Aesar
013209.06 |
5 g |
£195.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356658
|
Thermo Scientific Alfa Aesar
013209.09 |
10 g |
£356.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in various catalysts and catalytic reagents for organic synthesis, including the fabrication of various shapes of carbon nanostructures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
181266-82-0 | |
449.29 | |
YMTKZYSHNNXTIJ-UHFFFAOYSA-N | |
131675702 |
C15H21EuO6 | |
MFCD00192170 | |
europium iii 2,4-pentanedionate hydrate reo | |
[Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Specifications
181266-82-0 | |
C15H21EuO6 | |
Hygroscopic | |
Insoluble in water. | |
[Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
449.29 | |
449.29 (Anhydrous) | |
Powder | |
Europium(III) 2,4-pentanedionate hydrate |
140°C (decomposition) | |
MFCD00192170 | |
europium iii 2,4-pentanedionate hydrate reo | |
YMTKZYSHNNXTIJ-UHFFFAOYSA-N | |
europium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate | |
131675702 | |
99.9% | |
(REO) |
Safety and Handling
missing translation for 'einecsNumber' : 238-185-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only