missing translation for 'onlineSavingsMsg'
Learn More
Europium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11336668

Europium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11336668
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11336668

Brand: Thermo Scientific Alfa Aesar 013213.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula EuBr3·xH2O
MDL Number MFCD00049921

Specifications

Color White
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
Formula Weight 391.69 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Europium(III) bromide hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR18J012 Certificate TypeCertificate of Analysis Date24/05/2023 Catalog Number013213.14
Lot NumberR18J011 Certificate TypeCertificate of Analysis Date24/05/2023 Catalog Number013213.14
Lot NumberI08U028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013213.14,013213.06
Lot NumberZ04E008 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013213.14,013213.06
Lot NumberG05M18 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number013213.14,013213.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.