missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

£121.61 - £532.65

Chemical Identifiers

Molecular Formula EuBr3·xH2O
MDL Number MFCD00049921
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346668
View Documents
Thermo Scientific Alfa Aesar
013213.06
5 g
£121.61
5g
Please to purchase this item. Need a web account? Register with us today!
11336668
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
013213.14
25 g
£532.65
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

EuBr3·xH2O
MFCD00049921

Specifications

White
99.99% (REO)
MFCD00049921
391.69 (Anhydrous)
Hygroscopic
Crystals
EuBr3·xH2O
Soluble in water.
Odorless
Europium(III) bromide hydrate
Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'einecsNumber' : 237-349-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only