missing translation for 'onlineSavingsMsg'
Learn More
Europium(III) nitrate hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11397108

Europium(III) nitrate hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11397108
10 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11397108

Brand: Thermo Scientific Alfa Aesar 015290.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Europium(III) nitrate hexahydrate is used in the preparation of pharmaceuticals, agrochemicals and other chemical products. It is used as a catalyst and a component of specialized steels. It is also used in the synthesis of light emitting nanomaterials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-53-5
Molecular Formula EuH12N3O15
Molecular Weight (g/mol) 446.066
MDL Number MFCD00149700
InChI Key JVYYYCWKSSSCEI-UHFFFAOYSA-N
Synonym europium nitrate hexahydrate, europium iii nitrate, hexahydrate 1:3:6, nitric acid, europium 3+ salt, hexahydrate, eu.3no3.6h2o, ksc494m3l, europium 3+ trinitrate hexahydrate, europium iii nitrate hydrate, reacton, europium iii nitrate hexahydrate trace rare earth metals basis 1gShow MoreShow Less
PubChem CID 202256
IUPAC Name europium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Eu+3]

Specifications

Melting Point 85°C
Color Colorless
Physical Form Crystalline or Powder
Quantity 10 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 446.06 (337.97 Anhydrous)
Odor Odorless
Appearance Colorless crystalline
Chemical Name or Material Europium(III) nitrate hexahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU07K041 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog Number015290.18,015290.09,015290.04
Lot NumberM20I074 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog Number015290.18,015290.09,015290.04
Lot NumberY09G059 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog Number015290.18,015290.09,015290.04
Lot NumberJ30T009 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number015290.18,015290.09,015290.04
Lot NumberQ20C039 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number015290.18,015290.09,015290.04
compliance-icons
Product Identifier
  • Europium(III) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Europium(III) nitrate hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium(III) nitrate hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.