missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech CON 450 Handheld Meter Kit

Backlit, rugged waterproof meter is ideal for any environment

Brand:  Thermo Scientific™ 01X651633

478.47 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.


Product Code. 15397053

  • £718.42 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: Meter, conductivity cell (K = 1.0), Grip-Clip™, two AA batteries, hard carry case, USB cable, calibration solutions
This item is not returnable. View return policy

Description

Description

Backlit, rugged waterproof meter is ideal for any environment

  • Durable waterproof and dustproof design with IP67 rating    
  • Large, backlit custom display – View measurement with temperature and cell efficiency  
  • Grip-Clip™ – Quickly and easily secures the probe to your beaker or container    
  • Calibrate using automatically recognized standards or manually; to a single point across all ranges or to multiple points (one calibration point per each conductivity range)
  • Selectable cell constant – 0.10, 1.00, or 10.0  
  • Adjustable temperature coefficient from 0.00 to 10.00 % per ºC. Set to 0.0 % to meet USP methods for non-compensated measurements    
  • Adjustable conductivity-to-TDS factor – Adjustable 0.4 to 1.0    
  • Selectable ºC / ºF – Choose units to best fit your application    
  • Includes our easy to clean conductivity cell    
  • Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function    
  • Store up to 500 data sets with data – Use USB or RS232 output with optional data cable to send data to your software or our free communication software    
  • Built-in real-time clock stamps stored data and calibration data with time and date – Meets standards for GLP (Good Laboratory Practices)
  • Built-in stand with wall-mount – Allows for easy benchtop measurement or storage
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±1% Full-scale
Conductivity, TDS, Salinity, Temperature
0 to 200 mS
0 to 50 ppt
0.01 ppm to 0.1 ppt
-10°C to +110°C
±0.5°C (±0.9°F)
Stores up to 500 data sets
Meter: 7 7/8 x 3 1/8 x 2 1/4 in. (20 x 8.3 x 5.7 cm), Boxed: 1/8 x 9 1/8 x 2 3/4 in. (23 x 23 x 7 cm)
1 lb.
Two AA batteries (included), >300 hours (without backlight); AC adapter (optional)
USB, RS232
Auto/manual; 1 point/range
0.01 μS to 0.1 mS
0 to 200 ppt (TDS factor 1.0)
±1% Full-scale
0.01
Custom LCD with Backlight
500
0.5 kg
Eutech CON 450
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Brochures

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Eutech CON 450 Handheld Meter Kit >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.