Learn More
Thermo Scientific™ Eutech™ CyberScan PCD 650 Multi-parameter Meter
Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.
Brand: Thermo Scientific™ 01X430903
Description
Eutech™ CyberScan PCD 650 Multi-parameter Meter that measures pH, mV, ion, conductivity, tds, resistivity, salinity and dissolved oxygen. Combining the strengths of all CyberScan 600 series meters, the rugged CyberScan PCD650 is our top-of-the-line waterproof handheld that enables concurrent measurement of pH, conductivity, dissolved oxygen and temperature, all at the same time. Features a One-Glance-Tells-All Screen Display with Backlight Electrode status report, calibration data and simultaneous display of up to 4 parameters.
Features:
- Simultaneously measures and displays four parameters
- ±1% full scale accuracy at 3-decimal resolution across a wide measurement range
- Ion-measurement capabilities with 3-digit resolution
- Accepts two-cell and four-cell conductivity probe to measure up to 500 m/s — meter measures pure water!
- Selectable multi-point calibration for more accurate conductivity measurements or single-point calibration for quick, convenient calibration with no mistakes
- Measures 90.00mg/L in DO concentration and 600% in DO saturation
- Fast and easy DO calibration with 100% calibration option against atmospheric air or 0% calibration against a zero solution
- Salinity, Temperature and Pressure Compensation—accurate readings under varying conditions
- No warm up time required with galvanic system of measurement
- Easy-to-understand prompt messages for easy set-up, calibration and trouble-shooting
- Recorded readings individually stamped with time and date in GLP compliant format
- Auto-logging meter saves readings automatically within seconds to prevent data loss
- Wireless Information Technology: No wires, no cables—Transfer data from meter to computer with the press of a button
- Non-Volatile Memory protects information and meter settings, even when batteries run out
- Four Levels of Password Protection prevents tampering or accidental data deletion
Specifications
IrDA Wireless Communications | |
±1% FS + 1 LSD | |
±0.2 mV | |
0.1% ; ±0.2mg/L / ±0.2% | |
±1% FS +1 LSD | |
Automatic correction with built-in sensor | |
Up to 8 | |
0.010 to 10.000 | |
Multi-data Backlit Dot Matrix, 7.2 cm (H) X 5.4 cm (W) | |
pH, mV, Ion, Conductivity, TDS, Resistivity, Salinity, Dissolved Oxygen | |
±2000.0 mV | |
0 to 80 ppt | |
2 or 3 digits | |
-10.0°C to +110.0°C (Meter) | |
500 Data Sets with Date/Time stamping & Security password protection | |
450 to 825 mmHg / 59.9 to 109.9 kP | |
0 to 90.00 mg/L or ppm / 0 to 600% | |
0.1 mV | |
0.05% FS | |
Salinity Correction = Automatic, Resolution & Accuracy = 0.0 to 50.0ppt & 0.1 ppt | |
Automatic / Manual (0 to 100 °C); Temperature Coefficient = Linear & Pure | |
0.1 °C/°F |
0.1 mmHg or 0.1 kPa | |
±0.5% FS (mono); 1% FS (div) | |
±0.002 | |
±1% FS + 1 LSD | |
Calibration Due, HI-LO limit (out of range reading) | |
4 X 1.5 V AA Alkaline | |
1 (Offset) to 6 points | |
Waterproof CyberScan PCD 650 pH/ mV/ Ion/ Conductivity/ TDS/ Resistivity/ Salinity/ Dissolved Oxygen Meter with DAS software & multi probe holder. pH Buffer Options: USA, NIST, DIN, PWB, Custom | |
IP 67 | |
0 to 500 mS/cm | |
-2.000 to 20.000 | |
.. to 500.0 ppt | |
0.1/0.01/0.001 | |
Auto-Calibration; Data-logging options with time/date indexed, GLP compliance; Simultaneous Display and measurement of 4 parameters; Cal-due alarm; pH slope/offset Display & Self-diagnostic | |
9 V DC adapter, 500 mA | |
0.001 to 19900 | |
0.05% FS | |
0.01 mg/L or ppm | |
0.05% FS | |
±0.5°C, ±0.9°F | |
14.0°F to 230.0°F (Meter) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.