missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ CyberScan PCD 650 Multi-Parameter Meter
Click to view available options
Measurement Modes:
Acidity, Alkalinity, Temperature, Salinity
Unit Size:
Each
Description
Combining the strengths of all CyberScan 600 series meters, the CyberScan PCD650 is a top-of-the-line waterproof handheld that enables concurrent measurement of pH, conductivity, dissolved oxygen and temperature, all at the same time. Simultaneously measures and displays four parameters. For use in industrial, environmental, water quality testing, an aquaculture applications.
Features:
- ±1% full scale accuracy at 3-decimal resolution across a widemeasurement range
- Ion-measurement capabilities with 3-digit resolution
- Accepts two-cell and four-cell conductivity probe tomeasure up to 500m/s —metermeasures pure water
- Selectablemulti-point calibration formore accurate conductivitymeasurements or single-point calibration for quick, convenient calibration with nomistakes
- measures 90.00mg/L in DO concentration and 600% in DO saturation—one of the widest dissolved oxygenmeasurement ranges offered in the handheldmarket today
- Fast and easy DO calibration with 100% calibration option against atmospheric air or 0% calibration against a zero solution
- Salinity, Temperature and Pressure Compensation—accurate readings under varying conditions
- No warm up time required with galvanic system ofmeasurement
- One-Glance-Tells-All Screen Display with Backlight
- Electrode status report, calibration data and simultaneous display of up to 4 parameters
- Rugged and IP67-rated waterproof for applications in harsh environments.meter floats on water!
- Easy-to-understand promptmessages for easy set-up, calibration and trouble-shooting
- Recorded readings individually stamped with time and date in GLP compliant format
- Auto-loggingmeter saves readings automatically within seconds to prevent data loss
- HI-LO limit alarm for easy quality control –meter alerts when readings fall outside selected ranges
- Wireless Information Technology: No wires, no cables—Transfer data frommeter to computer with the press of a button
- Non-Volatilememory protects information andmeter settings, even when batteries run out
- Four Levels of Password Protection prevents tampering or accidental data deletion
- Calibration Alarm: Nomore out-dated calibration settings
Specifications
- pH Range: -2.000 to 20.000 pH
- of pH Calibration Points: 1 (Offset) to 6 points
- pH Buffer Options: USA, NIST, DIN, PWB, Custom
- mV Range: ±2000.0mV
- Ion Concentration Range: 001 to 19900
- DO Range: 0 to 90.00mg/L or ppm / 0 to 600%
- Salinity Correction: Automatic
- Barometric Pressure Correction: Automatic correction with built-in sensormmHg or 0.1 kPa
- Conductivity Range.. to 500mS/cm
- Cell Constant: 010 to 10.000
- Temperature Coefficient: Linear & Pure
- TDS Range: .. to 500.0 ppt
- Salinity Range 0 to 80 ppt
- Temperature Range: -10.0 to 110.0 °C / 14.0 to 230.0 °F (meter)
- Temperature Compensation: Automatic /manual (0 to 100 °C)
- memory: 500 Data Sets with Date/Time stamping & Security password protection
- Output: IrDA Wireless Communications
- Display:multi-data Back-lit dotmatrix; 7.2 cm (H) X 5.4 cm (W)
- Special Functions: Auto-Calibration; Data-logging options with time/date indexed, GLP compliance; Simultaneous Display andmeasurement of 4 parameters; Cal-due alarm; pH slope/offset Display & Self-diagnostic
- Power: 4 X 1.5 V 'AA' Alkor
Specifications
Specifications
| Outputs | IrDA Wireless Communication |
| Description | Waterproof CyberScan PCD 650 pH/ mV/ Ion/ Conductivity/ TDS/ Resistivity/ Salinity/ Dissolved Oxygen Meter with double junction pH electrode ECFC7252203B, 4-cell Conductivity probe CONSEN923J, Dissolved Oxygen probe ECDOHANDYNEW, DAS software, multi-probe holder, power adapter & carrying kit set (all probes comes with 3m cable). |
| Display Type | LCD Graphic, Backlit |
| IP Rating | IP67 |
| Measurement Modes | Acidity, Alkalinity, Temperature, Salinity |
| Range (Conductivity) | Up to 199.9 ÎĽS/cm, 200 ÎĽS/cm to 1999 ÎĽS/cm, 2 mS/cm to 20 mS/cm |
| Range (pH) | -2 to 20 |
| Range (Salinity) | 0 to 80 ppt |
| Temperature Range (Metric) | -10°C to +110°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Eutech™ CyberScan PCD 650 Multi-Parameter Meter >
Spot an opportunity for improvement?Share a Content Correction