Learn More
Thermo Scientific™ Eutech™ DO 6+ Dissolved Oxygen Meter ECDO601PLUSK
Eutech™ offers a wide range of Dissolved Oxygen (DO) meters and probes that measure the amount of gaseous oxygen dissolved in a solution in various educational, laboratory and industrial applications. 
Brand:  Thermo Scientific™ 01X370114
525.32 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Product Code. 11519224
  • £718.76 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Dissolved Oxygen (DO) meters and probes measure the amount of gaseous oxygen dissolved in a solution. In theory, the amount of DO in a solution is dependent on 3 factors, namely temperature, salinity and atmospheric pressure. The measurement of DO requires a special DO electrode that is made up of an anode, a cathode, electrolyte solution and a gas permeable membrane.
The Eutech™ DO 6+ Dissolved Oxygen Meter ECDO601PLUSK comes with 1m Cable Electrode ECDO6HANDY3M, 2 Assembled Membrane Cap Housing, 1 Refilling Electrolyte (10 ml) & Carrying Kit Set.
Features:
  • User Customisation for user's flexibility like toggling of various modes from ppm, mg/l to % saturation, view electrode diagnosis, etc.
  • Calibration can be performed at 100% and/or 0% and mg/l or ppm against a known solution
  • Automatic Temperature Compensation (ATC) guarantees the highest accuracy under varying temperature condition
  • Galvanic Cell allows instantaneous measurements, producing quick responses and eliminates polarisation; requires minimal maintenance
  • Automatically Compensates Salinity and Barometric Pressure after manual input for increased accuracy
  • Hold Function momentarily freezes reading for easy viewing
  •  Auto-Power Off saves battery power after non-use
  • Self-Diagnostic with message codes for easy troubleshooting
  • Large Custom LCD provides optimum viewing even at a distance
  • Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
  • Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Eutech DO 6+ Dissolved Oxygen Meter with 1m Cable Electrode ECDO6HANDY3M, 2 Assembled Membrane Cap Housing, 1 Refilling Electrolyte (10 ml) & Carrying Kit Set
500 to 1499 mmHg or 66.6 to 199.9 kPA
0.0°C to 50.0°C
± 0.3 °C (0.5 °F)
0 to 20.00 mg/l or ppm
0.1%
0.0 to 200/0 %
32°F to 122°F
4 x 1.4V batteries
Self-diagnostic; Electrode characteristics; Hold & Auto-Power Off after20 minutes
Up to 2 Calibration points; 100% in air and/or 0% in known solution
0.1°C (0.1°F)
LCD
Automatic correction after manual input
0.0 to 50.0 ppt
0.1%
> 700 hrs
± 1.5% Full Scale
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Eutech™ DO 6+ Dissolved Oxygen Meter ECDO601PLUSK >