Learn More
Thermo Scientific™ Eutech™ DO 6+ Dissolved Oxygen Meter
Eutech™ offers a wide range of Dissolved Oxygen (DO) meters and probes that measure the amount of gaseous oxygen dissolved in a solution in various educational, laboratory and industrial applications. 
Brand:  Thermo Scientific™ 01X370113
581.24 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 11509224
  • £760.29 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Dissolved Oxygen (DO) meters and probes measure the amount of gaseous oxygen dissolved in a solution. In theory, the amount of DO in a solution is dependent on 3 factors, namely temperature, salinity and atmospheric pressure. The measurement of DO requires a special DO electrode that is made up of an anode, a cathode, electrolyte solution and a gas permeable membrane.The Eutech™ DO 6+ Dissolved Oxygen Meter comes with 3m Cable Electrode ECDO6HANDY3M, 2 Assembled Membrane Cap Housing, 1 Refilling Electrolyte (10 ml) & Carrying Kit Set.
Features:
  • User Customisation for user's flexibility like toggling of variousmodes from ppm,mg/l to % saturation, view electrode diagnosis, etc.
  • Calibration can be performed at 100% and/or 0% andmg/l or ppm against a known solution
  • Automatic Temperature Compensation (ATC) guarantees the highest accuracy under varying temperature condition
  • Galvanic Cell allows instantaneousmeasurements, producing quick responses and eliminates polarisation; requiresminimalmaintenance
  • Automatically Compensates Salinity and Barometric Pressure aftermanual input for increased accuracy
  • Hold Functionmomentarily freezes reading for easy viewing
  • Auto-Power Off saves battery power after non-use
  • Self-Diagnostic withmessage codes for easy troubleshooting
  • Large Custom LCD provides optimum viewing even at a distance
  • Protective Rubber Boot shieldsmeter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
  • Rugged Carrying Case (Optional) provides handy on-sitemeter calibration andmeasurement
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Eutech™ DO 6+ Dissolved Oxygen Meter >