Learn More
Eutech Expert pH Pocket Tester
Perform on-the-go, fuss-free measurements for applications such as water treatment, environmental monitoring, education, hydroponics, agriculture, aquaculture, food & beverage and many more!
Brand: Eutech EXPERTPH
74.28 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.14852kg
Description
The Thermo Scientific™ Expert pH Pocket Tester is designed for on-the-go measurements of pH and temperature. The Expert pH Pocket Tester is best used in applications such as hydroponic gardening, aquaculture, agriculture, pools, cooling towers, and more!
- Large 4 digit double line display
- Simultaneous display of pH and temperature
- Battery life, ready, and calibration indicators
- Automatic temperature compensation
- Auto buffer recognition and calibration
- Multi language operational instructions
- Large 4 digit double line display and wide viewing angle make it easy to read and record measurements
- Battery life, ready, and calibration indicators
- Simultaneous display of pH with temperature
- Easy to operate and intuitive user interface with power, calibration, menu, and enter keys
- One push calibration: immerse sensor in calibration solution and press “Cal”
- Auto pH buffer recognition and calibration
- Gasketed and leak proof with an O-ring seal
- Transparent cap with a fill line indicator
- Symmetrical cap can be front or rear facing
- Cap secures to the top of tester when not in use
Additional features:
- Lanyard provision enables the user to easily transport the pocket tester for on-the-go measurements
- Pocket tester floats and meets Waterproof IP67 rating
- Wide, stable base allows the tester to independently stand vertically for measurements
- Automatic temperature compensation: adjusts readings according to temperature change for greater accuracy
- Auto-off function: pocket tester turns off after 8.5 minutes of non-use to extend battery life
- CD/RoHS Marking
- Multi language instructions available
Specifications
±0.1 pH | |
No | |
4 x 1.5V Micro Alkaline | |
1 to 3 | |
Expert pH pocket tester for measuring pH with temperature | |
IP 67 | |
0 to 14 | |
0.0°C to 60.0°C | |
5 to 85%, non-condensing | |
±0.6°C, ±1.0°F | |
32.0°F to 122.0°F | |
1 point | |
0.1°C, 0.1°F | |
6.7 in. | |
1.8 in. | |
1.2 in. | |
3.4 oz. |
±0.1 | |
100 hr. (average) | |
pH, temperature | |
CE, FCC Class A, ROHS-2 | |
4-digit, Double-line | |
pH, Temperature | |
0.1 | |
Expert pH tester with batteries | |
0.1pH | |
Yes | |
5°C to 45°C (ambient) | |
32.0°F to 140.0°F | |
1 Year | |
17 cm | |
4.5 cm | |
3 cm | |
96.3 g |