missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ Ion 2700 Meter
Quick and easy measurement with this ion-selective meter.
Brand: Thermo Scientific™ 01X543904
918.65 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Intuitive, self-diagnostic and flexible with advanced set-up options for user-customization, the Eutech ION 2700 series comes with a large, one-glance-sees-all screen. View pH, ION or Redox reading together with temperature, electrode status, calibration points, date and time all at once. All configurations include 30X427301 RS232 cable, 100/240 VAC power adapter and an integral electrode holder.
Features:
- Up to 6-point push button calibration with auto-buffer recognition
- Quick, easy electrode diagnosis with pH slope and offset display
- Non-volatile memory holds up to 500 data points – time and date-stamped for GLP compliance
- Bi-directional RS232 for easy data transfer to computer
- Cal-due alarm – no more out-dated calibrations!
- Auto-logging function for convenient continuous monitoring
- Limit alarm alerts when reading falls out of range
- Password protection for setup and calibration
- Buffer Sets: USA, NIST, DIN, User 1, User 2, Custom
Applications:
- Educational: Useful for most laboratory, ecological studies and other applications.
- Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing, where GLP data-management is required. Use in all types of food processing
Specifications
RS232 | |
±0.2 mV | |
Up to 6 preset or 5 custom | |
Up to 6 preset or 5 custom | |
pH, Ion, Redox | |
-2.000 to 20.000 | |
0.1/0.01/0.001 | |
Integral Electrode Holder & 100/240 VAC Adapter and RS232 cable | |
0.001 to 19999 ppm (±2000 mV) | |
±2000.0 mV | |
0.1 mV | |
ATC / MTC (0 to 100 °C) (pH only) | |
0.1 °C / 0.1 °F |
0.5% full scale (monovalent ion), 1% full scale (divalent ion) | |
±0.002 + 1 LSD | |
2 to 8 | |
Eutech ION 2700 Meter With pH Electrode (ECFG7370101B), ATC Probe (PH5TEMB01P) & pH Electrode Refill Solution, 60 mL | |
±2000.0 mV | |
2 or 3 digits | |
0.0°C to 100.0°C | |
9 V DC adapter, 1.3 A (100/240 VAC, SMPS) | |
±2000.0 mV | |
±2000.0 mV | |
±0.3°C, ±0.5°F | |
32.0°F to 212.0°F |