missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ Ion 2700 Meter

Quick and easy measurement with this ion-selective meter.

Brand:  Thermo Scientific™ 01X543903

 View more versions of this product

Product Code. 12903517

  • £1025.58 / Each
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Intuitive, self-diagnostic and flexible with advanced set-up options for user-customization, the Eutech ION 2700 series comes with a large, one-glance-sees-all screen. View pH, ION or Redox reading together with temperature, electrode status, calibration points, date and time all at once. All configurations include 30X427301 RS232 cable, 100/240 VAC power adapter and an integral electrode holder.

Features:

  • Up to 6-point push button calibration with auto-buffer recognition
  • Quick, easy electrode diagnosis with pH slope and offset display
  • Non-volatile memory holds up to 500 data points – time and date-stamped for GLP compliance
  • Bi-directional RS232 for easy data transfer to computer
  • Cal-due alarm – no more out-dated calibrations!
  • Auto-logging function for convenient continuous monitoring
  • Limit alarm alerts when reading falls out of range
  • Password protection for setup and calibration
  • Buffer Sets: USA, NIST, DIN, User 1, User 2, Custom

Applications:

  • Educational: Useful for most laboratory, ecological studies and other applications.
  • Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing, where GLP data-management is required. Use in all types of food processing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RS232
±0.2 mV
Up to 6 preset or 5 custom
Up to 6 preset or 5 custom
pH, Ion, Redox
-2.000 to 20.000
0.1/0.01/0.001
Integral Electrode Holder & 100/240 VAC Adapter and RS232 cable
0.001 to 19999 ppm (±2000 mV)
±2000.0 mV
0.1 mV
ATC / MTC (0 to 100 °C) (pH only)
0.1 °C / 0.1 °F
0.5% full scale (monovalent ion), 1% full scale (divalent ion)
±0.002 + 1 LSD
2 to 8
Eutech Ion 2700 Meter Only (includes Integral Electrode Holder & 100/240 VAC Adapter and RS232 cable)
±2000.0 mV
2 or 3 digits
0.0°C to 100.0°C
9 V DC adapter, 1.3 A (100/240 VAC, SMPS)
±2000.0 mV
±2000.0 mV
±0.3°C, ±0.5°F
32.0°F to 212.0°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers