Learn More
Thermo Scientific™ Eutech™ PC 450 Multi-parameter Meter Kit
Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.
Brand: Thermo Scientific™ 01X651640
726.72 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.96200kg
Description
Your all-in-one solution for multi-parameter research work in the lab. The multi-parameter Eutech PC 450 measures pH / mV / Conductivity / Total Dissolved Solids / Temperature. It accepts a single pH/Con probe or separate pH and conductivity probes.
Features
- Durable waterproof and dustproof design with IP67 rating
- Switch between pH,mV, conductivity and TDS with a press of a button
- Large, backlit custom display – Lets you viewmeasurement with temperature
- Grip-Clip™ – Quickly and easily secures separate pH and conductivity electrodes simultaneously to your sample container or beaker
- Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). Newmanual option allows calibration with up to 5 custom buffer standards you
- Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
- Calibration due alarm – Set a visible reminder when it is time to recalibrate themeter
- Intuitive, user-friendly icons – Electrode status displays pH slope or cell constant
- Auto-range conductivity from 0 to 199.9mS in five ranges – With up to five calibration points
- Selectable cell constant – 0.10, 1.00 or 10.0
- Adjustable temperature coefficient – From 0.00 to 10.00 % per °C
- Conductivity-to-TDS factor – Adjustable from 0.40 to 1.00
- Selectable °C/°F – Choose units to best fit your application
- Ready indicator,manual or automaticmeasurement hold feature and optional automatic shutoff function
- Store up to 500 data sets; easy USB or RS232 output with optional data cables
- Built-in real-time clock stamps stored data and calibration data with time and date –meets standards for GLP (Good Laboratory Practices)
- 500 hours battery life or use optional universal power adapter
- Built-in stand with wall-mount – Allows for easy benchtopmeasurement and storage
Specifications
Product Specification: Description
pH
- Range: -2.00 to 16.00 pH
- Resolution: 0.01 pH
- Accuracy: ±0.01 pH
- Calibration: Up to 6 points: USA, NIST, DIN or custom
mV
- Range: ±2000mV
- Resolution: 0.1mV ±999.9mV; 1mV beyond
- Accuracy: ±0.2mV +2 LSD or +0.5 % of reading; whichever is greater
- Calibration: ±200mV
Conductivity
- Range: 0 to 200mS; 0 to 20mS if using COmBI03BW probe
- Resolution: 0.01 μS to 0.1mS
- Accuracy: ±1 % full scale
- Calibration: Auto/manual; one point per range
TDS
- Range: 0 to 200 ppt (TDS factor 1.0)
- Resolution: 0.01 ppm to 0.1 ppt
- Accuracy: ±1 % full scale
- Calibration:manual; 1 point/range
Temperature
- Range: -10.0 to 110.0 °C (14.0 to 230.0 °F)
- Resolution: 0.1 °C/°F
- Accuracy: ±0.5 °C; ±0.9 °F
- Calibration: Offset in 0.1 increments, ±10 °C or ±18 °F
Others
- Display: Custom LCD with backlight
- memory: Stores up to 500 data sets
- Output: USB or RS232
- Real-Time Clock: Time and date stamp on calibration and stored data
- Temperature Compensation: Automatic ormanual from -10 to 110 °C
- Power: Two AA batteries (included), >100 hours (without backlight); AC adapter (optional)
- Dimensions (L x W x H):meter: 7 7/8 x 3 1/8 x 2 1/4 in. (20 x 8.3 x 5.7 cm), Boxed: 9 x 9 1/8 x 2 3/4 in. (23 x 23 x 7 cm
- Weight:meter: 1 lb (0.5 kg), Boxed: 1.4 lb (0.7 kg)
Specifications
USB, RS232 | |
LCD Graphic, Backlit | |
ORP, Salinity, Resistivity, pH, Conductivity | |
0 ppt to 200 ppt | |
-10°C to +110°C |
PC 450 Meter Kit: Meter, double-junction pH electrode, conductivity cell (K = 1.0), Grip-Clip™, two AA batteries, hard carry case, USB cable, calibration solutions | |
IP 67 | |
0 mS to 200 mS, 0 mS to 20 mS | |
0.01 |