Learn More
Thermo Scientific™ Eutech™ pH 150 Meter Kit
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Brand: Thermo Scientific™ 01X651609
446.89 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Eutech™ offers a wide range of pH meters and electrodes. pH 150 Meter Kit is an an IP67-rated waterproof pH meter that offers measurement of pH / mV / Temperature.
Features
- Large, three line LCD – View pH ormVmeasurement with temperature and % slope
- Built-inmemory function – Stores up to 150measurements along with temperature
- Auto buffer recognition – Automatically identifies USA, NIST or DIN buffer sets for rapid calibration
- Power with AA batteries or use optional AC adapter for benchtopmeasurements
- Calibration due alarm – Set a visible reminder when it is time to recalibrate themeter
- Selectable °F/°C – Choose units to best fit your application
- FeaturesmV offset – Calibrate to a Redox/ORP standard or use for pH electrode diagnostics
- Ready indicator,manual or automaticmeasurement hold feature and optional automatic shutoff function
- % slope and temperature displayed with eachmeasurement
- Built-in stand – Ideal for easy benchtop or long-term use
- Wall-mount option – Ideal to save on bench space
- Includes innovative Grip-Clip™ – Quickly and easily secures the electrode to your beaker or container
- Compatible with pH electrodes having a BNC connector
Specifications
Product Specification: Description
pH
- Range: -2.00 to 16.00 pH
- Resolution: 0.01 pH
- Accuracy: ±0.01 pH
- Calibration: Up to 5 points (USA & NIST), Up to 6 points (DIN)
mV
- Range: ±2000mV
- Resolution: 0.1mV ±999.9mV; 1mV beyond
- Accuracy: ±0.2mV +2 LSD or +0.5 % of reading; whichever is greater
- Calibration: ±200mV
Temperature
- Range: -10.0 to 110.0 °C (14.0 to 230.0 °F)
- Resolution: 0.1 °C/°F
- Accuracy: ±0.5 °C; ±0.9 °F
- Calibration: Offset in 0.1 increments, ±10 °C or ±18 °F
Others
- Display: Three line custom LCD
- Temperature Compensation: Automatic ormanual from -10 to 110 °C
- memory: Stores up to 150 data sets
- Power: Two AA batteries (included), >700 hours continuous use; AC adapter (optional)
- Dimensions (L x W x H):meter: 7 7/8 x 3 1/8 x 2 1/4 in. (20 x 8.3 x 5.7 cm), Boxed: 9 1/8 x 9 1/8 x 2 3/4 in. (23 x 23 x 7 cm)
- Weight:meter: 1 lb (0.5 kg), Boxed: 1.4 lb (0.7 kg)

Specifications
pH 150 Meter Kit: Meter with double-junction pH electrode ECFC7252201B and ATC probe PHWPTEM01W, Grip-Clip™, two AA batteries, hard carry case, pH buffers, storage solution, rinse solution | |
±0.2 or ±0.05%, whichever is greater | |
0.01 | |
-10°C to +110°C |
0.5°C/0.9°F | |
±0.01 | |
Up to 5 | |
LCD |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
28Hours
1Minutes
48Seconds
24
Your input is important to us. Please complete this form to provide feedback related to the content on this product.