Learn More
Eutech™ pH Buffers and Solutions, Color-Coded
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes
Brand: Thermo Scientific Eutech 01X211213
Code : NEW
Additional Details : Weight : 0.48600kg
Description
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.
Features:
- Buffers and calibration solutions packed in 60mL, 480 ml or 1L durable plastic bottles – economical and convenient for laboratory use.
Buffer Color Codes:
- Pink = pH 4.01
- Yellow = pH 7.00
- Blue = pH 10.01
Specifications
Yellow | |
pH Meter and Electrode | |
480 mL |
pH 7.00 Buffer, Yellow | |
7.00 | |
Buffer |