Learn More
Eutech™ pH Buffers and Solutions, Color-Coded
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes
£14.63 - £51.29
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
15432000
|
Thermo Scientific Eutech
01X639001 |
pH Buffer Kit, pH 4.01 (red), 7.00 (yellow), 10.01 (blue) | 480 mL Ea. |
£51.29
1 set |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15238728
|
Thermo Scientific Eutech
01X211233 |
pH 4.01 Buffer, Red | 1 L |
£21.00
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15122299
|
Thermo Scientific Eutech
01X211213 |
pH 7.00 Buffer, Yellow | 480 mL |
£14.71
480mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15248728
|
Thermo Scientific Eutech
01X211234 |
pH 7.00 Buffer, Yellow | 1 L |
£21.00
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15152299
|
Thermo Scientific Eutech
01X211205 |
pH 10.01 Buffer, Blue | 480 mL |
£14.63
480mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15258728
|
Thermo Scientific Eutech
01X211235 |
pH 10.01 Buffer, Blue | 1 L |
£20.91
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.
Features:
- Buffers and calibration solutions packed in 60mL, 480 ml or 1L durable plastic bottles – economical and convenient for laboratory use.
Buffer Color Codes:
- Pink = pH 4.01
- Yellow = pH 7.00
- Blue = pH 10.01
Specifications
pH Meter and Electrode |