Learn More
TDS 442 Standard Solution, Eutech™
For TDS calibration and standards, choose from our available Eutech™'s TDS Standards, Calibrations, and Solutions
Brand: Thermo Scientific Eutech 01X109101
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Eutech™ TDS Standard Solutions are used as standards for TDS meters. Get quick and accurate conductivity TDS measurements using Eutech™ TDS meters in the field, laboratories, and schools.
Features:
- Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
3000 ppm 442 Calibration Solution | |
480 mL | |
Colorless | |
7 | |
Laboratory Use |
Plastic Bottle | |
Standard Solution | |
TDS Standard | |
∼100°C | |
Liquid |