missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDS 442 Standard Solution, Eutech™
For TDS calibration and standards, choose from our available Eutech™'s TDS Standards, Calibrations, and Solutions
£14.20 - £14.63
Specifications
Packaging | Plastic Bottle |
---|---|
Quantity | 480 mL |
Solution Type | Standard Solution |
Color | Colorless |
Type | TDS Standard |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15107623
|
Thermo Scientific Eutech
01X109104 |
1000 ppm 442 Standard Solution |
£14.20
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15197613
|
Thermo Scientific Eutech
01X109102 |
300 ppm 442 Standard Solution |
£14.20
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15117623
|
Thermo Scientific Eutech
01X109101 |
3000 ppm 442 Calibration Solution |
£14.20
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11549124
|
Thermo Scientific Eutech
01X109105 |
50 ppm 442 Calibration Solution |
£14.63
500mL |
In Stock
Log in to see stock. |
|||||
Description
Eutech™ TDS Standard Solutions are used as standards for TDS meters. Get quick and accurate conductivity TDS measurements using Eutech™ TDS meters in the field, laboratories, and schools.
Features:
- Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
Plastic Bottle | |
Standard Solution | |
TDS Standard | |
Laboratory Use |
480 mL | |
Colorless | |
7 | |
Liquid |