Learn More
Heidolph™ Evaporating Flask
For use with rotary evaporators
Brand:  Heidolph™ 514-75000-00
Additional Details : Weight : 0.42000kg
 View more versions of this product
Product Code. 10545872
  • £168.00 / Each
Estimated Shipment: 11-09-2024
to see stock.
Add to basket
Description
Description
Evaporating flasks come with a standard NS 29/32 joint size but are also available in NS 24/29
Specifications
Specifications
Clear
Evaporating Flask
2000 mL
Glass
Rotary evaporators