Learn More
EVSCO™ Three-Way, Two-Position Elliptic Valve
Use these elliptic valves for liquid, gas and vacuum applications
Brand: EVSCO™ 98150-02
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Valves are easy to disassemble.
- Simply pull out the handle and push your stem through the bottom allowing you to clean and maintain valves in-line.
- Four-position valves are clearly marked with flow patterns to ensure correct handle position.
- Choose from FDA-grade PP or PVDF bodies.
- All valves have O-rings made of Viton, 316 SS handles, threaded connections and nonrestrictive orifice.
Specifications
Three-Way, Two-Position Elliptic Valve | |
Polypropylene, Viton | |
3/8 in. | |
3/8 in. NPT(F) |