Learn More
EVSCO™ Three-Way, Two-Position Elliptic Valve
Use these elliptic valves for liquid, gas and vacuum applications
£90.75 - £115.00
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11858083
|
EVSCO™
98150-02 |
3/8 in. |
£90.75
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11845312
|
EVSCO™
98151-01 |
1/4 in. |
£115.00
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
- Valves are easy to disassemble.
- Simply pull out the handle and push your stem through the bottom allowing you to clean and maintain valves in-line.
- Four-position valves are clearly marked with flow patterns to ensure correct handle position.
- Choose from FDA-grade PP or PVDF bodies.
- All valves have O-rings made of Viton, 316 SS handles, threaded connections and nonrestrictive orifice.
Specifications
Three-Way, Two-Position Elliptic Valve |