Learn More
SCAT Exhaust Filter, V3.0
The SCAT Exhaust Filter V3.0 for SafetyWasteCaps guarantees over 90% CTC adsorption for maximum air purity due to three integrated activated carbon types. This makes it the most powerful activated carbon filter medium on the HPLC laboratory market.
Brand: SCAT 490336
151.24 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.25800kg
Description
- Block off harmful vapours and ensure for safe pressure equalization in the waste containers
- 3-layer filter media with high performance spectrum for acids and alkalis
- Optimized for binding HPLC buffer solutions and absorbing organic solvent vapors
- The natural pore structure of the activated carbon prevents sticking or clogging
- Precise indication of the next replacement time by means of a time indicator
- Available in sizes (S/M/L) with different service lives (3/6/12 months)
- The use of exhaust filters can contribute to the reduction of the air exchange rate
Specifications
Multi-Pk. (Service Life 2 x 6 Months) | |
HPLC Systems | |
Exhaust Filter | |
SCAT Safety Waste Caps & Universal Waste Hub JAN |
Indicator | |
M | |
Polypropylene | |
GL14 |