missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT Exhaust Filter, V3.0
The SCAT Exhaust Filter V3.0 for SafetyWasteCaps guarantees over 90% CTC adsorption for maximum air purity due to three integrated activated carbon types. This makes it the most powerful activated carbon filter medium on the HPLC laboratory market.
£68.05 - £245.62
Specifications
For Use With (Application) | HPLC Systems |
---|---|
Material | Polypropylene |
Thread Size | GL14 |
Product Code | Brand | Size | Includes | Display Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Includes | Display Type | Price | Quantity & Availability | |||
17238870
|
SCAT
410534 |
S | Single Pk. (Service Life 3 Months) | Indicator |
£68.05
Each |
In Stock
Log in to see stock. |
|||
17258870
|
SCAT
490335 |
S | Multi-Pk. (Service Life 4 x 3 Months) | Indicator |
£245.62
Pack of 4 |
In Stock
Log in to see stock. |
|||
17218870
|
SCAT
407982 |
M | Single Pk. (Service Life 6 Months) | Label |
£101.00
Each |
In Stock
Log in to see stock. |
|||
17248870
|
SCAT
410535 |
M | Single Pk. (Service Life 6 Months) | Indicator |
£106.00
Each |
In Stock
Log in to see stock. |
|||
17278870
|
SCAT
490914 |
M | Multi-Pk. (Service Life 2 x 6 Months) | Label |
£196.08
Pack of 2 |
In Stock
Log in to see stock. |
|||
17268870
|
SCAT
490336 |
M | Multi-Pk. (Service Life 2 x 6 Months) | Indicator |
£200.21
Pack of 2 |
In Stock
Log in to see stock. |
|||
17228870
|
SCAT
407986 |
L | Single Pk. (Service Life 12 Months) | Indicator |
£143.00
Each |
In Stock
Log in to see stock. |
|||
17288870
|
SCAT
490986 |
L | Multi-Pk. (Service Life 2 x 12 Months) | Indicator |
£227.00
Pack of 2 |
In Stock
Log in to see stock. |
|||
17926970
|
SCAT™
407983 |
Large | Single Pk. (Service Life 12 Months) | Label |
£137.23
Each |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17936970
|
SCAT™
490915 |
Large | Multi-Pk. (Service Life 2 x 12 Months) | Label |
£218.14
Pack of 2 |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Block off harmful vapours and ensure for safe pressure equalization in the waste containers
- 3-layer filter media with high performance spectrum for acids and alkalis
- Optimized for binding HPLC buffer solutions and absorbing organic solvent vapors
- The natural pore structure of the activated carbon prevents sticking or clogging
- Precise indication of the next replacement time by means of a time indicator
- Available in sizes (S/M/L) with different service lives (3/6/12 months)
- The use of exhaust filters can contribute to the reduction of the air exchange rate
Specifications
HPLC Systems | |
Polypropylene | |
GL14 |