missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ GnTI- Expression System Kit

Expi293 GnTI- Expression System Kit is part of the Expi293 platform for rapid, high-yield transient protein expression from mammalian 293 cells

Brand:  Gibco™ A39250

Additional Details : Weight : 2.00000kg

Product Code. 16366395

  • £2310.00 / 1 set
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

System kit is centered on engineered Expi293F cells that do not have N-acetylglucosaminyltransferase I (GnTI) activity and therefore lack complex N-glycans. The cell line was established by the selective knockout of the GnTI gene in parental Expi293F cells. This cell line is a powerful tool for the high-yield expression of homogeneously glycosylated recombinant proteins.

Features:

  • Homogeneous N-glycosylation of expressed proteins
  • Equivalent protein yields to parental Expi293F cells - up to 1 g/L
  • Significantly higher protein yields than other HEK293 GnTI- cell line alternatives
  • Suspension, high-density culture of Expi293F GnTI- cells in Expi293 Expression Medium
  • High-efficiency transient transfection using ExpiFectamine 293 reagent in combination with specialized transfection enhancers

Components included:

  • 1 vial of frozen Expi293F GnTI- cells
  • 1 L of Expi293 Expression Medium
  • 1 ExpiFectamine 293 transfection kit sufficient to transfect 1 L of culture
  • 100 mL Opti-Plex Complexation Buffer
  • 1 PNGase F Glycan Cleavage kit
  • pRABBIT IgG IRES-EmGFP positive control vector

All components of the Expi293 GnTI- Expression System are also available for individual purchase

Specifications

Specifications

Transfection
Approved for shipment at Room Temperature or on Wet or Dry Ice
Expi293™
• Expi293F GnTI- Cells (1x107 cells/vial), store in vapor-phase liquid nitrogen
• 1 L Expi293 Expression Medium, store at 2–8°C (protect from light)
• ExpiFectamine 293 Transfection Kit for 1 L of culture, store at 2–8°C
• 100 mL Opti-Plex Complexation Buffer, store at 2–8°C (protect from light)
• PNGase F Glycan Cleavage Kit, store at -20°C
• pRABBIT IgG IRES-EmGFP Positive Control Vector, store at 2–8°C or long term storage at -20°C

Expi293F™
Protein Production
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.