missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiCHO™ Expression System Kit

Product Code. p-200089243
Click to view available options
Cell Line:
CHO-S, SFM Adapted
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15441923

Brand: Gibco™ A29133

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

2 vials frozen ExpiCHO-S™ cells, 1 bottle 1000mL ExpiCHO™ Expression Medium, 1 ExpiFectamine™ CHO transfection kit for 1 L of culture, 1 vial Antibody Expressing Positive Control Vector, 1 bottle 100mL OptiPRO™ SFM; store all components at +2° - +8° C, except frozen cells stored in liquid nitrogen

The ExpiCHO Expression System Kit combines the power of rapid, ultra-high-yield transient protein production in suspension culture with the benefit of expression in Chinese hamster ovary (CHO) cells.

The ExpiCHO Expression System Kit combines the power of rapid, ultra–high-yield transient protein production in suspension culture with the benefit of expression in Chinese hamster ovary (CHO) cells. It is based on high-density culture of ExpiCHO-S cells in ExpiCHO Expression Medium. Transient expression is powered by ExpiFectamine CHO transfection reagent, designed specifically for high-density CHO suspension culture, in combination with a specialized transfection enhancer and culture feed.

All components work together synergistically with expression yields of 1–3 g per L of transfected culture demonstrated for some antibody and non-antibody proteins.

The ExpiCHO Expression System enables:

  • Milligram to multigram yields of recombinant protein per liter of culture in 5–14 days post-transfection
  • Robust and reproducible transfection results, for greater confidence in results and ability to use CHO cells from research to discovery
  • Transfection efficiency of > 90% for high-density suspension CHO cell culture, with matching transfection enhancer and culture feed to boost transfection performance and protein expression
  • Use of one-third to one-half less plasmid DNA than other transfection reagents 
  • Flexible, reliable culture and transfection protocols with multiple options that readily fit into current transient expression workflows
  • Scalable expression for culture formats from  <1 mL in deep well plates up to 5 L when using Nalgene angled-bottom shake flasks

The components included in the ExpiCHO Expression System Kit are: 2 vials of frozen ExpiCHO-S cells, 1 L of ExpiCHO Expression Medium, 1 ExpiFectamine CHO Transfection Kit sufficient to transfect 1 L of culture, OptiPRO SFM, and an antibody-expressing positive control vector.

ExpiCHO-S cells, ExpiCHO Expression Medium, ExpiFectamine CHO Transfection Kit, and OptiPRO SFM may also be purchased separately.

All components of the system are animal origin–free.

TRUSTED_SUSTAINABILITY

Specifications

Type Expression System
Protein Tag Untagged
Delivery Type Transfection
Quantity 1 kit
Content And Storage • 2 vials frozen ExpiCHO-S cells
• 1000 mL ExpiCHO Expression Medium
• 1 ExpiFectamine CHO Transfection Kit for 1 L of culture
• 1 vial Antibody Expressing Positive Control Vector
• 100 mL OptiPRO SFM

Store all components at 2-8°C, except frozen cells which should be stored in liquid nitrogen.
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Cell Line CHO-S, SFM Adapted
Expression System Mammalian
Product Line ExpiCHO
For Use With (Application) Protein Production, Constitutive Expression
Promoter CMV
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt