Learn More
Gibco™ ExpiSf™ Protein Production Kit
Gibco™ ExpiSf™ Protein Production Kit is designed to significantly boost protein expression from high-density suspension cultures of Sf9 insect cells using the ExpiSf Expression System.
Brand:  Gibco™ A3767807
 View more versions of this product
Product Code. 15839126
  • £1230.00 / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Delivers greater than 80% more protein yield in high-density suspension Sf9 cell culture than ExpiSf CD Medium alone
  • Includes ExpiSf Enhancer, which is specifically designed to work in concert with ExpiSf CD Medium to significantly boost protein expression from baculovirus-infected insect cell cultures
  • Demonstrates direct scalability of baculovirus infection from culture volumes of 4mL in deep-well plates to >3 L in shake flasks
The ExpiSf Protein Production Kit is specifically designed to work with ExpiSf9 Cells (Cat. No. A35243) in the ExpiSf Expression System. The kit supplies sufficient ExpiSf Enhancer and ExpiSf CD Medium to infect 1 L, 10 L, or 50 L of culture. ExpiSf CD Medium is also available for purchase separately (Cat. No. A3767802).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Media and reagents, ExpiSf CD Meidum and ExpiSf Enhancer
10 L bag ExpiSf CD Medium, store in the dark at 2°C to 8°C; 40 mL ExpiSf Enhancer, store (at 2°C to 8°C; Shelf Life: 12 months from date of manufacture)
Insect Cells
Approved for shipment at Room Temperature or on Wet Ice
Baculovirus
10 L (ExpiSf CD Medium), 40 mL (ExpiSf Enhancer)
Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free
Liquid
Cell-Based Expression
ExpiSf™