Learn More
Memmert™ Extended Overtemperature Protection for UF260
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand:  Memmert™ UF260A6
Product Code. 17589105
  • £166.00 / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
Specifications
UF260 Oven
Integrated Pt100 Sensor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Extended Overtemperature Protection for UF260 > For Use With: UF260