missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Extension Rod
Buerkle™ Extension Rod makes it possible to take samples from various depths.
Brand: Buerkle™ 5380-1006
80.23 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Seven different drill bits are available, each of which is designed for a particular soil type
- For various applications
Specifications
Extension Rod | |
Autoclavable | |
Silver | |
For Sampling Very Small Quantities of Highly Potent, Expensive or Active Powders for Analysis | |
1000 mm |
Stainless Steel (V4A) | |
Equipment for PharmaPicker | |
Electro-polished | |
PharmaPicker |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction