Learn More
Buerkle™ Extension Rod
Buerkle™ Extension Rod makes it possible to take samples from various depths.
Brand:  Buerkle™ 5336-0100
45.97 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10123152
  • £51.65 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Seven different drill bits are available, each of which is designed for a particular soil type
  • For various applications
Specifications
Specifications
Extension Rod
Autoclavable
Gray
1000 mm
Polypropylene
Equipment for ChemoSampler
ChemoSampler