Learn More
Buerkle™ Extension Rod
Buerkle™ Extension Rod makes it possible to take samples from various depths.
Brand: Buerkle™ 5389-0001
This item is not returnable.
View return policy
Description
- Seven different drill bits are available, each of which is designed for a particular soil type
- For various applications
Specifications
Extension Rod | |
Autoclavable | |
Pure White | |
600 mm |
Steel Core Encapsulated in PTFE | |
Equipment for PTFE scoop | |
PTFE Scoop |