Learn More
Ismatec™ Extension Tubing, Tygon ST
For use with 2- and 3-stop color-coded tubing.
Brand:  Ismatec™ SC0044T
 View more versions of this product
Product Code. 10027331
  • £163.00 / 10m
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Inexpensive, all-around tubing for general laboratory applications.
  • Transparent
  • Resistant to almost all inorganic chemicals
  • Tasteless
  • Smooth, polished inner wall
  • Low gas permeability
  • Non-aging and non-oxidizing
  • High dialectric constant
Specifications
Specifications
FDA 21 CFR175.300
1.85 mm
10 m
−50°C
Acids: Good, Alkaline solutions: Good, Solvents: Not rec., Pressure: Fair, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited
Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C (250°F); tubing will appear milky.Gas sterilization with Ethylene oxide.Not recommended for sterilization with radiation.
74°C
Autoclavable
Tygon ST R-3603, R-3607
General laboratory applications, 2- and 3-stop color-coded tubing.
1 Reel
I.D. 1.85 mm
Transparent