Learn More
Thermo Scientific™ Extensor Long PCR Enzyme Mix Products
Amplify long PCR products up to 20kb with this synergistically-acting PCR enzyme blend of Taq DNA polymerase and a proprietary proofreading enzyme.
£538.00 - £550.34
Specifications
Content And Storage | Store at -20°C. Avoid freeze/thaw cycles |
---|---|
For Use With (Application) | Cloning |
Concentration | 2X |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
11819512
|
Thermo Scientific™
AB-0792/B |
Extensor Master Mix (Buffer 1 Formulation) | 500 25µL Rxns | ||||||
12340130
|
Thermo Scientific™
AB-0794/B |
Extensor ReddyMix Master Mix (Buffer 1 Formulation) | 400 x 25µL Rxns | ||||||
Description
Extensor Long Range PCR Enzyme is a blend of ThermoPrime Taq DNA Polymerase and a proprietary thermostable proofreading enzyme. The two enzymes act synergistically to generate long PCR products, up to 20 kb. Extensor Long Range Enzyme includes 2 buffer formulations; Buffer 1 for templates up to 12 kb in length, and Buffer 2 for templates 12 to 20 kb and GC-rich or other difficult templates. This enzyme blend is also available in 2X Master Mix and ReddyMix formulations. The ReddyMix Master Mix contains an inert red dye and a density reagent that allows for direct loading onto agarose gels.
Highlights
- Proprietary enzyme blend amplifies targets up to 20 kb in length
- Alternative buffering system for success with GC rich targets
Specifications
Store at -20°C. Avoid freeze/thaw cycles | |
Cloning | |
2X |
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.