missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 87-950 (ex EXTRA)

Extra protection for heavy duty applications.

Brand:  Ansell™ 87950080BP

34.22 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12475120

  • £47.60 / 12 pairs
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • 50% thicker than standard flocklined gloves.
  • Extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats.
  • 100% natural rubber with no fillers. Excellent resistance to mechanical hazards plus increased chemical protection.
  • Chlorinated. For better grip and increased chemical resistance.
  • 100% cotton flocklining. Makes a heavy glove softer and more comfortable to wear. Helps absorb perspiration.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rubber Latex
Natural Rubber Latex
No
8
0.75 mm
Straight
144Pack
87-950
EN388: 2003, EN374:2003, EN388:2016, EN ISO 374-1:2016, EN ISO 374-5:2016
Natural Rubber Latex
Immersion Gloves
87-950
Black
Cotton Flocking
Many ketones, salts, detergents, alcohols, alkalis and fats
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ AlphaTec™ 87-950 (ex EXTRA) > Size: 8

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.