missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ 87-950 (ex EXTRA)
Extra protection for heavy duty applications.
£31.10 - £49.27
Specifications
Back Material | Rubber Latex |
---|---|
Certifications/Compliance | EN388: 2003, EN374:2003, EN388:2016, EN ISO 374-1:2016, EN ISO 374-5:2016 |
Cuff Material | Natural Rubber Latex |
Coating Material | Natural Rubber Latex |
Product Type | Immersion Gloves |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
12465120
|
Ansell™
87950070 |
7.5 | N/A | |||||||
12475120
|
Ansell™
87950080BP |
8 |
£47.60
12 pairs |
In Stock
Log in to see stock. |
||||||
12485120
|
Ansell™
87950090BP |
9 |
£49.27
12 pairs |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
12495120
|
Ansell™
87950100BP |
10 |
£49.27
12 pairs |
In Stock
Log in to see stock. |
||||||
12627041
|
Ansell™
87950110BP |
11 |
£47.60
12 pairs |
In Stock
Log in to see stock. |
||||||
Description
- 50% thicker than standard flocklined gloves.
- Extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats.
- 100% natural rubber with no fillers. Excellent resistance to mechanical hazards plus increased chemical protection.
- Chlorinated. For better grip and increased chemical resistance.
- 100% cotton flocklining. Makes a heavy glove softer and more comfortable to wear. Helps absorb perspiration.
Specifications
Rubber Latex | |
Natural Rubber Latex | |
Immersion Gloves | |
0.75 mm | |
Cotton Flocking | |
87-950 |
EN388: 2003, EN374:2003, EN388:2016, EN ISO 374-1:2016, EN ISO 374-5:2016 | |
Natural Rubber Latex | |
87-950 | |
Straight | |
Many ketones, salts, detergents, alcohols, alkalis and fats |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell™ AlphaTec™ 87-950 (ex EXTRA)