Learn More
Lenz Laborglasintrumente Extractors Acc. to Knöfler-Böhm
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05375043
Additional Details : Weight : 0.50000kg
Description
- The complete apparatuses are consisting of: Jacketed extractor head made of DURAN™ tubing, Dimroth condenser (details on page 71)
- With thread GL 14
- Made of DURAN™ tubing
- Round bottom flask
Specifications
150 mL | |
Duran Glass | |
NS 45/40 |