Learn More
Thermo Scientific™ EZ-Link™ Maleimide-PEG2-Biotin, No-Weigh™ Format
Biotinylate reduced cysteines with this short, maleimide-activated, sulfhydryl-reactive biotinylation reagent containing a 2-unit PEG spacer arm to enhance water solubility.
Brand: Thermo Scientific™ A39261
235.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Additional Details : Weight : 0.05200kg
Description
Thermo Scientific™ EZ-Link Maleimide-PEG2-Biotin is a mid-length, maleimide-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.
- Mid-length, maleimide-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.
- Protein labeling - biotinylate antibodies or other proteins for use in protein methods
- Thiol-reactive - reacts with sulfhydryls (-SH), such as the side-chain of cysteine (C)
- Maleimide-activated - perform reactions at pH 6.5 to 7.5 in buffers such as PBS
- Pegylated - spacer arm contains a hydrophilic, 2-unit, polyethylene glycol (PEG) group
- Enhances solubility - pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
- Irreversible - forms permanent thioether bonds; spacer arm cannot be cleaved
- Solubility - can be dissolved directly in aqueous buffers for labeling reactions
- Medium length - spacer arm (total length added to target) is 29.1 angstroms
Specifications
Maleimide-PEG2-Biotin | |
Store desiccated at 4°C. | |
Qualified for Ambient or Wet Ice | |
Cell-Impermeant | |
Maleimide | |
Biotin |
10 x 2 mg | |
Chemical Labeling | |
EZ-Link™ | |
Thiol | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Pegylated, 29.1Å |
For Research Use Only. Not for use in diagnostic procedures.